Learn More
Bio X Cell Antibody, anti-mouse TIM-1 (CD365), 100 MG
Supplier: Bio X Cell BE0314R100MG
The 3D10 monoclonal antibody reacts with mouse T cell immunoglobulin and mucin domain 1 (TIM-1), also known as CD365. It is preferentially expressed on TH2 cells and acts as a stimulatory molecule for T cell activation.
- Target: Mouse TIM-1 (CD365)
- Clone: 3D10
- Isotype: Rat IgG1, κ
- Purity: ≥95%
- Endotoxin level: ≤1 EU/mg
- Production: Purified in an animal-free facility
- Storage: Store at 4°C, do not freeze
- Applications: In vivo and in vitro TIM-1 blockade
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.