Learn More
Bio X Cell Bio X Cell, Mouse TIM-1 (CD365), Clone 3D10, 25 MG
Supplier: Bio X Cell BE0314R025MG
The 3D10 monoclonal antibody reacts with mouse T cell immunoglobulin and mucin domain 1 (TIM-1/CD365), a type I cell-surface glycoprotein that stimulates T cell activation and regulates immune responses. This antibody has been shown to block TIM-1 in vivo and enhance atherosclerosis in mouse studies.
- Clone: 3D10
- Reactivities: mouse
- Isotype: rat IgG1, κ
- Purification: purified from cell culture supernatant in an animal-free facility by Protein G. Purity: ≥95% (determined by SDS-PAGE).
- Formulation: PBS, pH 7.0. Contains no stabilizers or preservatives. Endotoxin: ≤1EU/mg (≤0.001EU/μg), determined by LAL assay.
- Reported applications: in vivo TIM-1 blockade, in vitro TIM-1 blockade
- Molecular weight: 150 kDa
- Storage: store at the stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.