Learn More
Bio X Cell InVivoMAb, Anti-Mouse TIM-1, 3D10, IgG1, 5 MG
Supplier: Bio X Cell BE0314R005MG
The 3D10 monoclonal antibody reacts with mouse T cell immunoglobulin and mucin domain 1 (TIM-1), also known as CD365. It is preferentially expressed on TH2 cells and has been identified as a stimulatory molecule for T cell activation. The 3D10 antibody has been shown to block TIM-1 in vivo and enhance atherosclerosis in mice studies.
- Applications: In vivo and in vitro TIM-1 blockade
- Host species: Rat
- Target species: Mouse
- Clone: 3D10
- Isotype: Rat IgG1, κ
- Purification method: Protein G
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Storage: Store at 4°C, do not freeze
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Molecular weight: 150 kDa
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.