Learn More
Bio X Cell Antibody, Anti-mouse TIM-1 (CD365), Clone 3D10, Rat IgG1, 50 MG
Supplier: Bio X Cell BE0314R050MG
This monoclonal antibody reacts with mouse T cell immunoglobulin and mucin domain 1 (TIM-1), also known as CD365. TIM-1 is a type I cell-surface glycoprotein involved in regulating the immune response, allergic responses, asthma, and transplant tolerance. The antibody blocks TIM-1 activity in vivo and in vitro.
- Clone: 3D10
- Host: Rat
- Isotype: IgG1,
- Reactivity: Mouse
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin level: 1 EU/mg
- Purity: 95% by SDS-PAGE
- Sterility: 0.2 m filtration
- Purified by Protein G
- Molecular weight: 150 kDa
- Applications: In vivo and in vitro TIM-1 blockade
- Storage: Store at 4C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.