Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Anti-human CD19, Clone 4G7, IgG1, 1 MG

Catalog No. 501528461
Encompass_Preferred

The 4G7 monoclonal antibody targets human CD19, a B cell-specific 95 kDa transmembrane glycoprotein. CD19 regulates B-cell receptor signaling and is highly expressed in lymphomas and leukemias, making it a valuable target for immunotherapy.

  • Clone: 4G7
  • Target: Human CD19
  • Host species: Mouse
  • Isotype: Mouse IgG1
  • Conjugation: Unconjugated
  • Formulation: PBS, pH 7.0; no stabilizers or preservatives
  • Endotoxin: ≤1 EU/mg
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Production: Animal-free facility
  • Purification: Protein G
  • Molecular weight: 150 kDa
  • Storage: Store at 4°C; do not freeze
  • Reported applications: Flow cytometry, functional assays, immunofluorescence, chimeric antigen receptor construction

Catalog No. 50-152-8461 Supplier Bio X Cell Supplier No. BE0281R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 4G7 monoclonal antibody targets human CD19, a B cell-specific 95 kDa transmembrane glycoprotein. CD19 regulates B-cell receptor signaling and is highly expressed in lymphomas and leukemias, making it a valuable target for immunotherapy.

  • Clone: 4G7
  • Target: Human CD19
  • Host species: Mouse
  • Isotype: Mouse IgG1
  • Conjugation: Unconjugated
  • Formulation: PBS, pH 7.0; no stabilizers or preservatives
  • Endotoxin: ≤1 EU/mg
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Production: Animal-free facility
  • Purification: Protein G
  • Molecular weight: 150 kDa
  • Storage: Store at 4°C; do not freeze
  • Reported applications: Flow cytometry, functional assays, immunofluorescence, chimeric antigen receptor construction

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.