Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell PSGL-1, Clone 4RA10, Rat IgG1, 1 MG

Catalog No. 501528385
Encompass_Preferred

This monoclonal antibody targets mouse P-selectin glycoprotein ligand-1 (PSGL-1), also known as CD162. PSGL-1 is a 230 kDa glycoprotein involved in leukocyte rolling, migration into inflamed tissues, and responses to vascular injury. The antibody is reported to block the binding of mouse leukocytes to CD62P and CD62L.

  • Host species: rat
  • Isotype: rat IgG1, κ
  • Immunogen: mouse PSGL-1 human IgG1 fusion protein
  • Purification method: protein G
  • Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
  • Endotoxin level: ≤1EU/mg (≤0.001EU/μg)
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Storage: store at stock concentration at 4°C, do not freeze
  • Reported applications: in vivo PSGL-1 blockade, immunohistochemistry (frozen)

Catalog No. 50-152-8385 Supplier Bio X Cell Supplier No. BE0186R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This monoclonal antibody targets mouse P-selectin glycoprotein ligand-1 (PSGL-1), also known as CD162. PSGL-1 is a 230 kDa glycoprotein involved in leukocyte rolling, migration into inflamed tissues, and responses to vascular injury. The antibody is reported to block the binding of mouse leukocytes to CD62P and CD62L.

  • Host species: rat
  • Isotype: rat IgG1, κ
  • Immunogen: mouse PSGL-1 human IgG1 fusion protein
  • Purification method: protein G
  • Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
  • Endotoxin level: ≤1EU/mg (≤0.001EU/μg)
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Storage: store at stock concentration at 4°C, do not freeze
  • Reported applications: in vivo PSGL-1 blockade, immunohistochemistry (frozen)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.