Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Bio X Cell, Anti-human EGFR Antibody, Clone 528, 1 MG

Catalog No. 501528459
Encompass_Preferred

The 528 monoclonal antibody reacts with human EGFR (epidermal growth factor receptor), also known as ErbB-1. This antibody has been reported to block EGF binding to its receptor and inhibit A431 tumor formation in nude mice.

  • Isotype: Mouse IgG2a
  • Immunogen: Purified EGFR from A431 cells
  • Reported applications: In vitro and in vivo EGFR blockade, Western blot, functional assays, immunoprecipitation, immunohistochemistry, immunofluorescence, flow cytometry
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Molecular weight: 150 kDa
  • Storage: Store undiluted at 4°C, protected from prolonged light exposure. Do not freeze.

Catalog No. 50-152-8459 Supplier Bio X Cell Supplier No. BE0279R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 528 monoclonal antibody reacts with human EGFR (epidermal growth factor receptor), also known as ErbB-1. This antibody has been reported to block EGF binding to its receptor and inhibit A431 tumor formation in nude mice.

  • Isotype: Mouse IgG2a
  • Immunogen: Purified EGFR from A431 cells
  • Reported applications: In vitro and in vivo EGFR blockade, Western blot, functional assays, immunoprecipitation, immunohistochemistry, immunofluorescence, flow cytometry
  • Purity: ≥95%
  • Sterility: 0.2 μm filtration
  • Molecular weight: 150 kDa
  • Storage: Store undiluted at 4°C, protected from prolonged light exposure. Do not freeze.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.