Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Antibody, Monoclonal, Rat IgG1, Mouse CSF1, In Vivo Neutralization, 1 MG

Catalog No. 501528401
Encompass_Preferred

The 5A1 monoclonal antibody reacts with mouse colony stimulating factor 1 (CSF1), also known as macrophage colony-stimulating factor (M-CSF). CSF1 is a hematopoietic growth factor expressed by various cell types, playing a role in regulating the proliferation, differentiation, and survival of monocytes, macrophages, and bone marrow progenitor cells. The 5A1 antibody has been shown to neutralize CSF1 in vivo.

  • Monoclonal antibody, clone 5A1
  • Isotype rat IgG1,
  • Reacts with mouse CSF1
  • Recommended for in vivo CSF1 neutralization
  • Formulated in PBS, pH 8.0, without stabilizers or preservatives
  • Purity greater than 95%
  • Endotoxin level less than or equal to 1 EU/mg
  • 0.2 m filtered for sterility
  • Produced in an animal-free facility
  • Protein G purified
  • Molecular weight 150 kDa
  • Store at 4C, do not freeze

Catalog No. 50-152-8401 Supplier Bio X Cell Supplier No. BE0204R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The 5A1 monoclonal antibody reacts with mouse colony stimulating factor 1 (CSF1), also known as macrophage colony-stimulating factor (M-CSF). CSF1 is a hematopoietic growth factor expressed by various cell types, playing a role in regulating the proliferation, differentiation, and survival of monocytes, macrophages, and bone marrow progenitor cells. The 5A1 antibody has been shown to neutralize CSF1 in vivo.

  • Monoclonal antibody, clone 5A1
  • Isotype rat IgG1,
  • Reacts with mouse CSF1
  • Recommended for in vivo CSF1 neutralization
  • Formulated in PBS, pH 8.0, without stabilizers or preservatives
  • Purity greater than 95%
  • Endotoxin level less than or equal to 1 EU/mg
  • 0.2 m filtered for sterility
  • Produced in an animal-free facility
  • Protein G purified
  • Molecular weight 150 kDa
  • Store at 4C, do not freeze

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.