Learn More
Bio X Cell InVivoMAb Antibody, Anti-mouse TL1A 5G4.6, 100 MG
Supplier: Bio X Cell BE0323R100MG
This monoclonal antibody targets the TNF family member TL1A (TNFSF15), found on activated T cells, dendritic cells, monocytes, and endothelial cells. It blocks TL1A binding to DR3, reducing disease severity in mouse models of colitis where autoreactive T cells are involved.
- Isotype: Armenian hamster IgG
- Purification method: Protein A
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Endotoxin level: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Storage: Store at 4°C, do not freeze
- Applications: in vivo TL1A neutralization, flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.