Learn More
Bio X Cell InVivoMAb anti-mouse TL1A, Clone 5G4.6, 25 MG
Supplier: Bio X Cell BE0323R025MG
The 5G4.6 monoclonal antibody targets the TNF family member TL1A (TNFSF15), which promotes effector T cell proliferation. This antibody blocks TL1A-DR3 interactions, reducing pathology in animal models involving autoreactive T cells.
- Isotype: Armenian hamster IgG
- Reacts with mouse TL1A (TNFSF15)
- Applications: In vivo TL1A neutralization, flow cytometry
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Purified from cell culture supernatant in an animal-free facility
- Purification method: Protein A
- Molecular weight: 150 kDa
- Storage: Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.