Learn More
Bio X Cell InVivoMAb Antibody, anti-mouse TL1A (TNFSF15), Armenian Hamster IgG, 25 MG
Supplier: Bio X Cell BE0323R005MG
This 5G4.6 monoclonal antibody targets TL1A (TNFSF15), a TNF family member expressed on various immune cells and induced by pro-inflammatory stimuli. Its receptor, DR3 (TNFRSF25), found on T cells, NK cells, and NKT cells, mediates T cell proliferation at inflammation sites. Blocking TL1A-DR3 interactions has shown reduced pathology in animal models of autoimmune diseases.
- Isotype: Armenian hamster IgG
- Immunogen: Recombinant murine TL1A
- Applications: in vivo TL1A neutralization, flow cytometry
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: 1 EU/mg
- Purity: 95% by SDS-PAGE
- Sterility: 0.2 m filtration
- Molecular weight: 150 kDa
- Storage: 4C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.