Learn More
Bio X Cell Antibody, CEACAM 1/3/5/6/8, Clone 6G5j, 1 MG
Supplier: Bio X Cell BE0362R001MG
The 6G5j monoclonal antibody targets human carcinoembryonic antigen-related cell adhesion molecule (CEACAM) 1, 3, 5, 6, and 8 (CD66a, d, e, c, and b). These highly glycosylated proteins are involved in cell adhesion, proliferation, differentiation, and immune cell activation.
- Clone: 6G5j
- Target: CEACAM 1, 3, 5, 6, and 8 (CD66a, d, e, c, and b)
- Host: Mouse
- Isotype: Mouse IgG1, κ
- Applications: Western blot, ELISA, immunofluorescence, immunohistochemistry (paraffin), flow cytometry
- Purification: Protein G, purified from cell culture supernatant in an animal-free facility
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Storage: Store at stock concentration at 4°C; do not freeze
- Endotoxin: ≤1 EU/mg (≤0.001 EU/µg)
- Purity: ≥95%
- Sterility: 0.2 µm filtration
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.