Learn More
Bio X Cell CEACAM 1/3/5/6/8, Clone 6G5j, IgG1, 25 MG
Supplier: Bio X Cell BE0362R025MG
The 6G5j monoclonal antibody reacts with an antigen epitope shared by mouse carcinoembryonic antigen-related cell adhesion molecule (CEACAM) 1, 3, 5, 6, and 8, also known as CD66a, d, e, c, and b. These CEACAMs are highly glycosylated membrane proteins involved in cell adhesion, proliferation, differentiation, apoptosis, and T and B cell activation.
- Reacts with CEACAM 1, 3, 5, 6, and 8
- Purified via Protein G from cell culture supernatant in an animal-free facility
- Purity ≥95% determined by SDS-PAGE
- Suitable for Western blot, ELISA, immunofluorescence, immunohistochemistry (paraffin), and flow cytometry
- Formulated in PBS, pH 7.0, with no stabilizers or preservatives
- Storage at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.