Learn More
Bio X Cell Monoclonal Antibody, Human CEACAM 6G5j, 50 MG
Supplier: Bio X Cell BE0362R050MG
The 6G5j monoclonal antibody reacts with an antigen epitope shared by human carcinoembryonic antigen-related cell adhesion molecule (CEACAM) 1, 3, 5, 6, and 8. These highly glycosylated membrane proteins are involved in cell adhesion, proliferation, differentiation, apoptosis, T and B cell activation, and serve as bacterial pathogen receptors (except for CEACAM 8).
- Isotype: Mouse IgG1, κ
- Immunogen: Human CEACAM1-Fc fusion protein
- Applications: Western blot, ELISA, Immunofluorescence, Immunohistochemistry (paraffin), Flow cytometry
- Purity: ≥95% determined by SDS-PAGE
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Purification: Protein G, 0.2 μm filtration from animal-free facility
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.