Learn More
Bio X Cell InVivoMAb, 6S2-19-4, Mouse CCR3, IgG2b, 25 MG
Supplier: Bio X Cell BE0316R025MG
The 6S2-19-4 monoclonal antibody reacts with mouse CCR3 (CD193), a G protein-coupled, seven transmembrane, chemokine receptor expressed on various hematopoietic cells. It selectively depletes eosinophils when administered in vivo, playing a role in atopic diseases and parasitic infection.
- Reacts with mouse CCR3 (CD193)
- Expressed on eosinophils, basophils, mast cells, and other hematopoietic cells
- Ligands include RANTES, eotaxin, eotaxin-3, MCP-3, and MIP1α
- Formulated in PBS, pH 7.0; contains no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Purified from cell culture supernatant in an animal-free facility
- Molecular weight: 150 kDa
- Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.