Learn More
Bio X Cell InVivoMAb Anti-Mouse CCR3, Clone 6S2-19-4, 5 MG
Supplier: Bio X Cell BE0316R005MG
The 6S2-19-4 monoclonal antibody reacts with mouse CCR3 (CD193), a G protein-coupled receptor highly expressed on eosinophils. This antibody selectively depletes eosinophils when administered in vivo, making it suitable for studies on atopic diseases and parasitic infections.
- Selectively depletes eosinophils in vivo
- Purified by Protein G from animal-free facility
- Formulation in PBS, pH 7.0, without stabilizers or preservatives
- Purity ≥95% as determined by SDS-PAGE
- Endotoxin level ≤1EU/mg
- Sterility by 0.2 μm filtration
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.