Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoMAb Antibody, Human CD25, 7G7B6, 1 MG

Catalog No. 501528240
Encompass_Preferred

The 7G7B6 monoclonal antibody reacts with human IL-2Rα (CD25), the 55 kDa ligand-binding subunit of the interleukin 2 receptor alpha chain. It is expressed on activated mature T and B lymphocytes, thymocyte subsets, pre-B cells, and T regulatory cells, playing roles in lymphocyte differentiation, activation, and proliferation.

  • Clone: 7G7B6
  • Reactivity: Human CD25 (IL-2Rα)
  • Isotype: Mouse IgG2a
  • Immunogen: Human PBMCs
  • Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
  • Purity: ≥95% by SDS-PAGE
  • Endotoxin: ≤1EU/mg
  • Sterility: 0.2 µm filtration
  • Purification method: Protein G
  • Storage: 4°C; do not freeze
  • Applications: Regulatory T cell depletion in humanized mice, immunoprecipitation, immunofluorescence

Catalog No. 50-152-8240 Supplier Bio X Cell Supplier No. BE0014R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 7G7B6 monoclonal antibody reacts with human IL-2Rα (CD25), the 55 kDa ligand-binding subunit of the interleukin 2 receptor alpha chain. It is expressed on activated mature T and B lymphocytes, thymocyte subsets, pre-B cells, and T regulatory cells, playing roles in lymphocyte differentiation, activation, and proliferation.

  • Clone: 7G7B6
  • Reactivity: Human CD25 (IL-2Rα)
  • Isotype: Mouse IgG2a
  • Immunogen: Human PBMCs
  • Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
  • Purity: ≥95% by SDS-PAGE
  • Endotoxin: ≤1EU/mg
  • Sterility: 0.2 µm filtration
  • Purification method: Protein G
  • Storage: 4°C; do not freeze
  • Applications: Regulatory T cell depletion in humanized mice, immunoprecipitation, immunofluorescence

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.