Learn More
Bio X Cell Antibody, Anti-Mouse CD71, Clone 8D3, 1 MG
Supplier: Bio X Cell BE0329R001MG
The 8D3 monoclonal antibody reacts with mouse CD71 (TfR1), a 170-180 kDa type II homodimeric transmembrane glycoprotein expressed on proliferating cells, reticulocytes, and erythroid precursors. It is crucial for cellular proliferation and iron import, and its high expression on malignant cells makes it an attractive target for drug delivery. The 8D3 antibody depletes CD71+ cells in vivo.
- Host: Rat
- Isotype: Rat IgG2a
- Reported applications: in vivo depletion of CD71+ cells, immunofluorescence, immunohistochemistry (frozen), western blot
- Purification: Protein G
- Formulation: PBS, pH 7.0. Contains no stabilizers or preservatives.
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.