Learn More
Bio X Cell Anti-Human CD28, Clone 9.3, IgG2a, 1 MG
Supplier: Bio X Cell BE0248R001MG
The 9.3 monoclonal antibody reacts with human CD28, a 45 kDa costimulatory receptor, expressed by thymocytes, peripheral T cells, and NK cells. Signaling through CD28 augments IL-2 and IL-2 receptor expression as well as cytotoxicity of CD3-activated T cells.
- Reacts with human CD28.
- Isotype mouse IgG2a.
- Immunogen human T lymphocytes.
- Purified by protein G.
- Formulated in PBS, pH 7.0, with no stabilizers or preservatives.
- Endotoxin ≤1EU/mg determined by LAL assay.
- Purity ≥95% determined by SDS-PAGE.
- Sterility 0.2 μm filtered.
- Produced in an animal-free facility.
- Recommended storage at 4°C, do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.