Learn More
Bio X Cell Antibody, C-Myc, 9E10, IgG1, 1 MG
Supplier: Bio X Cell BE0238R001MG
The 9E10 monoclonal antibody reacts with human c-myc, a 62 kDa transcription factor involved in cell cycle progression, apoptosis, and cellular transformation. Amplification of the c-myc gene is found in several human cancers, including lung, breast, and colon carcinomas. C-Myc is often added to proteins of interest using recombinant DNA technology, allowing the c-myc tag to be recognized by an antibody in various assays.
- Purified from cell culture supernatant in an animal-free facility
- High purity: ≥95% determined by SDS-PAGE
- Low endotoxin: ≤1 EU/mg (≤0.001 EU/µg) determined by LAL assay
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Sterility: 0.2 µm filtration
- Purification: Protein G
- Unconjugated, custom conjugation services available
- For in vivo applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.