Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Mouse IgG1 Antibody, T15 IgM, 1 MG

Catalog No. 501528298
Encompass_Preferred

The AB1-2 monoclonal antibody reacts with mouse IgM of the T15 idiotype, raised against purified immunoglobulins from MOPC 460, MOPC 5558, and HOPC 8 myelomas. This unconjugated antibody is purified from cell culture supernatant in an animal-free facility using Protein G, with a molecular weight of 150 kDa.

  • Reacts with mouse IgM of the T15 idiotype
  • Clone: AB1-2 (HB33)
  • Isotype: Mouse IgG1, κ
  • Molecular weight: 150 kDa
  • Purity: ≥95% (SDS-PAGE)
  • Endotoxin: ≤1EU/mg
  • Formulation: PBS, pH 7.0, no stabilizers or preservatives
  • Sterility: 0.2 μm filtration
  • Storage: 4°C, do not freeze

Catalog No. 50-152-8298 Supplier Bio X Cell Supplier No. BE0072R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The AB1-2 monoclonal antibody reacts with mouse IgM of the T15 idiotype, raised against purified immunoglobulins from MOPC 460, MOPC 5558, and HOPC 8 myelomas. This unconjugated antibody is purified from cell culture supernatant in an animal-free facility using Protein G, with a molecular weight of 150 kDa.

  • Reacts with mouse IgM of the T15 idiotype
  • Clone: AB1-2 (HB33)
  • Isotype: Mouse IgG1, κ
  • Molecular weight: 150 kDa
  • Purity: ≥95% (SDS-PAGE)
  • Endotoxin: ≤1EU/mg
  • Formulation: PBS, pH 7.0, no stabilizers or preservatives
  • Sterility: 0.2 μm filtration
  • Storage: 4°C, do not freeze

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.