Learn More
Bio X Cell anti-mouse IL-15, Clone AIO.3, 50 MG
Supplier: Bio X Cell BE0315R050MG
The AIO.3 monoclonal antibody reacts with mouse IL-15. IL-15 is a pro-inflammatory cytokine primarily produced by dendritic cells, epithelial cells, fibroblasts, and monocytes, playing crucial roles in the immune response. It stimulates proliferation of activated T cells, NK cells, and B cells, and induces antibody production. The AIO.3 antibody neutralizes IL-15 bioactivity in vitro and in vivo.
- Clone: AIO.3
- Reactivities: Mouse
- Isotype: Rat IgG2a, λ
- Immunogen: Recombinant mouse IL-15
- Reported applications: in vivo IL-15 neutralization, in vitro IL-15 neutralization
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin: ≤1EU/mg
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.