Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Antibody, Anti-mouse IL-1 alpha, ALF-161, 1 MG

Catalog No. 501528431
Encompass_Preferred

The ALF-161 monoclonal antibody reacts with precursor, secreted, and membrane-associated forms of mouse IL-1α, neutralizing its bioactivity. IL-1α is a 17 kDa pro-inflammatory cytokine.

  • Clone: ALF-161
  • Reactivities: Mouse
  • Isotype: Armenian hamster IgG
  • Purity: ≥95% (determined by SDS-PAGE)
  • Endotoxin: ≤1EU/mg (≤001EU/µg)
  • Sterility: 0.2 µm filtration
  • Production: Purified from cell culture supernatant (animal-free facility)
  • Purification: Protein A
  • Molecular Weight: 150 kDa
  • Storage: Store at stock concentration at 4°C; do not freeze
  • Reported applications: in vivo IL-1α neutralization, in vitro IL-1α neutralization
  • Immunogen: Recombinant mouse IL-1α
  • Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)

Catalog No. 50-152-8431 Supplier Bio X Cell Supplier No. BE0243R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The ALF-161 monoclonal antibody reacts with precursor, secreted, and membrane-associated forms of mouse IL-1α, neutralizing its bioactivity. IL-1α is a 17 kDa pro-inflammatory cytokine.

  • Clone: ALF-161
  • Reactivities: Mouse
  • Isotype: Armenian hamster IgG
  • Purity: ≥95% (determined by SDS-PAGE)
  • Endotoxin: ≤1EU/mg (≤001EU/µg)
  • Sterility: 0.2 µm filtration
  • Production: Purified from cell culture supernatant (animal-free facility)
  • Purification: Protein A
  • Molecular Weight: 150 kDa
  • Storage: Store at stock concentration at 4°C; do not freeze
  • Reported applications: in vivo IL-1α neutralization, in vitro IL-1α neutralization
  • Immunogen: Recombinant mouse IL-1α
  • Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.