Learn More
Bio X Cell InVivoMAb, Clone B122, anti-IL-1 beta, 25 MG
Catalog No. 501528435
Encompass_Preferred
The InVivoMAb anti-mouse/rat IL-1β is a high-quality monoclonal antibody designed for in vivo and in vitro research applications. It targets precursor and mature secreted forms of mouse and rat Interleukin-1 beta (IL-1β), a 17 kDa pro-inflammatory cytokine. The B122 clone effectively neutralizes the bioactivity of natural or recombinant IL-1β, making it an invaluable tool for studying inflammatory processes and immune responses.
  • Effectively neutralizes natural and recombinant mouse/rat IL-1β
  • Reacts with mouse and rat IL-1β for versatile comparative studies
  • Suitable for in vivo/in vitro neutralization studies and ELISA
  • High purity (≥95%) and low endotoxin (≤1EU/mg) for sensitive in vivo use
  • Animal-free production reduces contamination and ensures lot consistency
  • Extensively cited (110+ citations) for proven efficacy
Catalog No. 50-152-8435 Supplier Bio X Cell Supplier No. BE0246R025MG
The InVivoMAb anti-mouse/rat IL-1β is a high-quality monoclonal antibody designed for in vivo and in vitro research applications. It targets precursor and mature secreted forms of mouse and rat Interleukin-1 beta (IL-1β), a 17 kDa pro-inflammatory cytokine. The B122 clone effectively neutralizes the bioactivity of natural or recombinant IL-1β, making it an invaluable tool for studying inflammatory processes and immune responses.
  • Effectively neutralizes natural and recombinant mouse/rat IL-1β
  • Reacts with mouse and rat IL-1β for versatile comparative studies
  • Suitable for in vivo/in vitro neutralization studies and ELISA
  • High purity (≥95%) and low endotoxin (≤1EU/mg) for sensitive in vivo use
  • Animal-free production reduces contamination and ensures lot consistency
  • Extensively cited (110+ citations) for proven efficacy