Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoMAb anti-mouse/rat IL-1 beta, B122, 50 MG

Catalog No. NC2892928
Encompass_Preferred

The B122 monoclonal antibody reacts with precursor and mature secreted forms of mouse and rat IL-1β, a 17 kDa pro-inflammatory cytokine primarily produced by monocytes. This antibody neutralizes the bioactivity of natural or recombinant IL-1β.

  • Isotype: Armenian hamster IgG
  • Immunogen: Recombinant mouse IL-1β
  • Reported applications: in vivo IL-1β neutralization, in vitro IL-1β neutralization, ELISA
  • Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Purity: ≥95% determined by SDS-PAGE
  • Sterility: 0.2 μm filtered
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein G
  • Molecular weight: 150 kDa
  • Storage: Store at stock concentration at 4°C. Do not freeze.

Catalog No. NC2892928 Supplier Bio X Cell Supplier No. BE0246R050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The B122 monoclonal antibody reacts with precursor and mature secreted forms of mouse and rat IL-1β, a 17 kDa pro-inflammatory cytokine primarily produced by monocytes. This antibody neutralizes the bioactivity of natural or recombinant IL-1β.

  • Isotype: Armenian hamster IgG
  • Immunogen: Recombinant mouse IL-1β
  • Reported applications: in vivo IL-1β neutralization, in vitro IL-1β neutralization, ELISA
  • Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Purity: ≥95% determined by SDS-PAGE
  • Sterility: 0.2 μm filtered
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Purification: Protein G
  • Molecular weight: 150 kDa
  • Storage: Store at stock concentration at 4°C. Do not freeze.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.