Learn More
Bio X Cell InVivoMAb anti-human CD71, Clone B3/25, 5 MG
Supplier: Bio X Cell BE0367R005MG
The Bio X Cell InVivoMAb anti-human CD71 (TfR1) antibody, Clone B3/25, is a mouse IgG1 monoclonal antibody engineered for various research applications including immunofluorescence and neutralization/blocking studies.
- Target antigen: CD71 (TfR1) – human
- Host species: Mouse
- Isotype: IgG1
- Clonality: Monoclonal
- Applications: Immunofluorescence, in vitro neutralization and blocking, in vivo neutralization and blocking
- Concentration: 4–11 mg/mL
- Purity: ≥95% (determined by SDS-PAGE)
- Storage conditions: 2°C to 8°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.