Learn More
Bio X Cell anti-human CD71, B3/25, Mouse, 50 MG
Supplier: Bio X Cell BE0367R050MG
This product is an InVivoMAb anti-human CD71 (TfR1) antibody, clone B3/25, developed by Bio X Cell. It is suitable for applications such as ImmunoFluorescence and is reactive with human samples.
- Suitable for ImmunoFluorescence and other applications
- Reactive with human samples
- Intended for research use only
- Concentration 4–11 mg/mL
- Estimated purity ≥95% by SDS-PAGE
- Store at 2°C to 8°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.