Learn More
Bio X Cell Monoclonal Antibody, mouse TIM-3, B8.2C12, 1 MG
Supplier: Bio X Cell BE0275R001MG
The B8.2C12 monoclonal antibody targets mouse TIM-3 (T cell immunoglobulin and mucin domain-3), also known as CD366. It strongly binds to the BALB/c allele of TIM-3, with weaker reactivity to the C57Bl/6 allele. TIM-3 is a 60 kDa immune checkpoint receptor, crucial for inducing tolerance and investigated as a target for cancer immunotherapy.
- Designed for in vivo functional studies
- Immune checkpoint regulator, targeting TIM-3
- Purity of ≥95% determined by SDS-PAGE
- Endotoxin of ≤1EU/mg determined by LAL assay
- Sterile with 0.2 μm filtration
- Produced in an animal-free facility
- Recommended isotype control: InVivoMAb rat IgG1, anti-horseradish peroxidase
- Recommended dilution buffer: InVivoPure pH 7.0 Dilution Buffer
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.