Learn More
Bio X Cell Antibody, Anti-Human CTLA-4, BN13, IgG2a, 1 MG
Supplier: Bio X Cell BE0190R001MG
The BN13 monoclonal antibody targets human CTLA-4 (CD152), a 33 kDa cell surface receptor expressed on activated T and B lymphocytes. This antibody negatively regulates cell-mediated immune responses and is utilized as an immune checkpoint blockade target for cancer treatment.
- Isotype: Mouse IgG2a, κ
- Conjugation: Unconjugated
- Immunogen: Human CTLA-4 recombinant protein
- Reported applications: In vitro CTLA-4 neutralization, Flow cytometry
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%; determined by SDS-PAGE
- Sterility: 0.2 μm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.