Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Mouse IgG2a Isotype Control, C1.18.4, 1 MG

Catalog No. 501528305
Encompass_Preferred

The C1.18.4 monoclonal antibody serves as a non-reactive isotype-matched control for mouse IgG2a antibodies across various in vivo and in vitro applications.

  • Isotype: Mouse IgG2a, κ
  • Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg), determined by LAL assay
  • Purity: ≥95%, determined by SDS-PAGE
  • Sterility: 0.2 μm filtration
  • Purification: Protein G
  • Molecular weight: 150 kDa
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Storage: Store at 4°C at stock concentration; do not freeze

Catalog No. 50-152-8305 Supplier Bio X Cell Supplier No. BE0085R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The C1.18.4 monoclonal antibody serves as a non-reactive isotype-matched control for mouse IgG2a antibodies across various in vivo and in vitro applications.

  • Isotype: Mouse IgG2a, κ
  • Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg), determined by LAL assay
  • Purity: ≥95%, determined by SDS-PAGE
  • Sterility: 0.2 μm filtration
  • Purification: Protein G
  • Molecular weight: 150 kDa
  • Production: Purified from cell culture supernatant in an animal-free facility
  • Storage: Store at 4°C at stock concentration; do not freeze

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.