Learn More
Bio X Cell InVivoMAb, Anti-Mouse IL-12 p35, 25 MG
Supplier: Bio X Cell BE0371R025MG
The C18.2 monoclonal antibody reacts with mouse interleukin-12 (IL-12) p35, a 70 kDa cytokine secreted by activated monocytes, macrophages, and dendritic cells. IL-12 regulates cell-mediated immune responses and plays a key role in developing Th1 responses, leading to IFNγ and IL-2 production.
- Clone: C18.2
- Reactivity: Mouse
- Isotype: Rat IgG2a, κ
- Unconjugated
- Immunogen: Recombinant mouse IL-12 p70
- Endotoxin: ≤1EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Reported applications: in vivo and in vitro IL-12 p35 neutralization, ELISA, immunoprecipitation
- Purification method: Protein G
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Storage: Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.