Learn More
Bio X Cell Anti-ICOS (CD278), Clone C398.4A, Unconjugated, 1 MG
Supplier: Bio X Cell BE0353R001MG
This monoclonal antibody reacts with mouse, human, rat, and monkey ICOS (CD278), a 47-57 kDa glycoprotein on activated T cells. It co-stimulates T and B cell responses and is used for in vitro T cell activation.
- Reactivities: mouse, human, rat, monkey
- Isotype: Armenian hamster IgG
- Clonality: monoclonal, clone C398.4A
- Conjugation: unconjugated
- Immunogen: mouse T cell line D10.G4.1
- Formulation: PBS, pH 7.0, no stabilizers/preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Purification: protein A
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
- Applications: in vitro T cell stimulation, flow cytometry, immunoprecipitation
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.