Learn More
Bio X Cell Monoclonal Antibody, Clone C398.4A, Anti-ICOS (CD278), Armenian Hamster, 100 MG, 100 MG
Supplier: Bio X Cell BE0353R100MG
This monoclonal antibody reacts with mouse, human, rat, and monkey ICOS (inducible T cell co-stimulator), also known as CD278. ICOS is a 47-57 kDa homodimeric glycoprotein belonging to the CD28 family of costimulatory molecules, expressed on activated T cells. It co-stimulates T and B cell responses upon ICOSL binding, which is expressed on antigen-presenting cells. ICOS signaling is also important for maintaining regulatory T cell homeostasis. This antibody is commonly used to co-stimulate T cell activation and proliferation in vitro and does not block the binding of ICOS to ICOSL.
- Clone C398.4A
- Target antigen: ICOS (CD278)
- Reactivity with mouse, human, rat, and monkey ICOS
- Host species: Armenian hamster
- Isotype: Armenian hamster IgG
- Applications include in vitro T cell stimulation/activation, flow cytometry, and immunoprecipitation
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Purity of 95%, d
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.