Learn More
Bio X Cell InVivoMAb Antibody, anti-ICOS C398.4A, Unconjugated, 25 MG
Supplier: Bio X Cell BE0353R025MG
The C398.4A monoclonal antibody targets ICOS (CD278), an inducible T cell co-stimulator expressed on activated T cells. This antibody is commonly used for in vitro T cell activation and proliferation, co-stimulating T and B cell responses without blocking ICOS-ICOSL binding.
- Isotype: Armenian hamster IgG
- Unconjugated format for various applications
- Targets ICOS (CD278) from mouse, human, rat, and monkey
- Applications include in vitro T cell stimulation/activation, flow cytometry, and immunoprecipitation
- Purified from cell culture supernatant in an animal-free facility
- High purity (≥95%) and low endotoxin (≤1EU/mg, ≤0.001EU/μg)
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.