Learn More
Bio X Cell Monoclonal Antibody, ICOS (CD278), C398.4A, 5 MG
Supplier: Bio X Cell BE0353R005MG
This C398.4A monoclonal antibody targets ICOS (CD278), an inducible T cell co-stimulator, found on activated T cells. It reacts with mouse, human, rat, and monkey ICOS, co-stimulating T and B cell responses upon ICOSL binding. It is commonly used for in vitro T cell activation and proliferation.
- Reacts with mouse, human, rat, and monkey ICOS (CD278)
- Clone C398.4A
- Host species: Armenian hamster
- Isotype: Armenian hamster IgG
- Purified by Protein A
- Applications: in vitro T cell stimulation/activation, flow cytometry, immunoprecipitation
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin ≤1 EU/mg; purity ≥95%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.