Learn More
Bio X Cell InVivoMAb, anti-human CD2, CB.219, IgG2b, 1 MG
Supplier: Bio X Cell BE0354R001MG
The InVivoMAb anti-human CD2 monoclonal antibody, clone CB.219, targets human CD2, a 45-58 kD type I transmembrane glycoprotein involved in T cell and NK cell activation and regulation of cytolytic activities.
- Reacts with human CD2
- Purified by protein A
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin ≤1EU/mg (≤0.001EU/μg)
- Sterilized by 0.2 μm filtration
- Purity ≥95% by SDS-PAGE
- Recommended storage at 4°C; do not freeze
- Applications include in vivo CD2 blockade in huCD2tg mice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.