Learn More
Bio X Cell Antibody, Clone CB.219, Anti-Human CD2, 25 MG
Supplier: Bio X Cell BE0354R025MG
The CB.219 monoclonal antibody reacts with human CD2, a transmembrane glycoprotein (45-58 kD). CD2, also known as LFA-2, T11, or Ly37, is expressed on peripheral blood lymphocytes, thymocytes, T cells, and NK cells, functioning as an adhesion receptor binding to CD58. The antibody interacts with the N-terminal CD58-binding region of CD2.
- Isotype: mouse IgG2b, κ
- Application: in vivo CD2 blockade in huCD2tg mice
- Unconjugated
- Immunogen: human T4+ CTL clone
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: from animal-free facility
- Purification: protein A
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.