Learn More
Bio X Cell Monoclonal Antibody, Human CD2, CB.219, 5 MG
Supplier: Bio X Cell BE0354R005MG
The CB.219 monoclonal antibody reacts with human CD2, a 45-58 kD type I transmembrane glycoprotein. CD2 acts as an adhesion receptor, binding to CD58 to activate CD2-positive T cells and NK cells. This antibody interacts with the N-terminal CD58-binding region of CD2.
- Conjugation: Unconjugated
- Immunogen: Human T4+ CTL clone
- Applications: in vivo CD2 blockade in huCD2tg mice
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg (≤0.001 EU/µg)
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 µm filtration
- Production: Purified from animal-free cell culture supernatant
- Purification method: Protein A
- Molecular weight: 150 kDa
- Storage: Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.