Learn More
Waters Corp mAb Charge Variant Standard, Unique Mix of Intact Proteins
Catalog No. NC1781045 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This standard is specifically designed to help benchmark charge variant chromatography when using the BioResolve SCX columns. ;It contains a unique mix of intact proteins relevant to Biopharma customers: trytophan, conalbumin and the NIST RM 8671 (NIST mAb). ;This standard is also used to QC each batch of BioResolve SCX mAb material.
Catalog No. NC1781045 Supplier Waters Corp Supplier No. 186009065
This standard is specifically designed to help benchmark charge variant chromatography when using the BioResolve SCX columns. ;It contains a unique mix of intact proteins relevant to Biopharma customers: trytophan, conalbumin and the NIST RM 8671 (NIST mAb). ;This standard is also used to QC each batch of BioResolve SCX mAb material.