Learn More
Cygnus Technologies M220 mAb Coated Microplate, 96 Wells, Anti-MVM-MVP Coating, Unit of Measure: 1 Plate
Catalog No. NC3501103
Encompass
The Cygnus Technologies M220 mAb Coated Microplate is a specialized 96-well plate ideal for the MockV MVM Kit Immuno-qPCR assay. With its pre-coated surface featuring anti-MVM-MVP, this microplate streamlines the capture and detection of target analytes, enhancing assay performance. Its design specifically aids researchers in achieving reliable results, making it a superior choice for those conducting immuno-qPCR assays.
Catalog No. NC3501103 Supplier Cygnus Technologies Supplier No. M220
The Cygnus Technologies M220 mAb Coated Microplate is a specialized 96-well plate ideal for the MockV MVM Kit Immuno-qPCR assay. With its pre-coated surface featuring anti-MVM-MVP, this microplate streamlines the capture and detection of target analytes, enhancing assay performance. Its design specifically aids researchers in achieving reliable results, making it a superior choice for those conducting immuno-qPCR assays.