Learn More
Bio X Cell Anti-Mouse NKG2D CX5, Rat IgG1, 1 MG
Supplier: Bio X Cell BE0334R001MG
The CX5 monoclonal antibody reacts with mouse NKG2D, a type II transmembrane lectin-like glycoprotein (CD314). It is expressed on NK cells, NKT cells, CD8 T cells, γ/δ T cells, and macrophages. This non-depleting antibody blocks NKG2D binding to its ligands and mediates receptor internalization.
- For in vivo and in vitro NKG2D blockade
- Validated for flow cytometry
- Purified by Protein G from cell culture supernatant in an animal-free facility
- Isotype: Rat IgG1, κ
- Purity: ≥95% determined by SDS-PAGE
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.