Learn More
Bio X Cell Monoclonal Antibody, anti-mouse CD28, High-Purity, 1 MG
Supplier: Bio X Cell BE0328R001MG
The D665 monoclonal antibody reacts with mouse CD28, a 45 kDa costimulatory receptor, commonly used to induce Treg cell expansion in vivo in mouse models of disease. It stimulates IL-2 and IL-2 receptor expression and T cell proliferation.
- Reacts with mouse CD28
- Clone: D665
- Isotype: Mouse IgG1, κ
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: Animal-free facility
- Purification: Protein G
- Molecular Weight: 150 kDa
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.