Learn More
Bio X Cell Antibody, D665, Mouse CD28, IgG1, 5 MG
Supplier: Bio X Cell BE0328R005MG
This D665 monoclonal antibody reacts with mouse CD28, a 45 kDa costimulatory receptor. It is expressed by thymocytes, most peripheral T cells, and NK cells and commonly used to induce the expansion of Treg cells in vivo in various mouse models.
- Reacts with mouse CD28, a 45 kDa costimulatory receptor
- Reported applications: in vivo T cell stimulation/activation, in vitro T cell stimulation/activation
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: purified from cell culture supernatant in an animal-free facility
- Purification: protein G
- Molecular weight: 150 kDa
- Storage: store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.