Learn More
Bio X Cell Monoclonal Antibody, Mouse E-Cadherin, DECMA-1, 100 MG
Supplier: Bio X Cell BE0352R100MG
The DECMA-1 monoclonal antibody reacts with the extracellular domain of mouse E-Cadherin, a 120-kDa transmembrane cell-cell adhesion glycoprotein also known as CD324 and uvomorulin. E-Cadherin is a critical regulator of epithelial junction formation, involved in development, bacterial pathogenesis, and tumor invasion.
- Host species: Rat
- Isotype: IgG1, κ
- Purity: ≥95% by SDS-PAGE
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Purification: Protein G from animal-free facility
- Applications: In vivo/in vitro E-Cadherin neutralization, immunofluorescence, immunoprecipitation, Western blot
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.