Learn More
Bio X Cell Antibody, E-Cadherin, DECMA-1, Rat IgG1, 25 MG
Supplier: Bio X Cell BE0352R025MG
The DECMA-1 monoclonal antibody reacts with the extracellular domain of mouse E-Cadherin (CD324), a 120-kDa transmembrane cell-cell adhesion glycoprotein crucial for epithelial junction formation. It is involved in development, bacterial pathogenesis, and tumor invasion.
- Reacts with extracellular domain of mouse E-Cadherin (CD324)
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% (determined by SDS-PAGE)
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Reported applications: in vivo E-Cadherin neutralization, in vitro E-Cadherin neutralization, Immunofluorescence, Immunoprecipitation, Western blot
- Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.