Learn More
Bio X Cell Antibody, DECMA-1, E-Cadherin, Rat IgG1, 5 MG
Supplier: Bio X Cell BE0352R005MG
The DECMA-1 monoclonal antibody reacts with the extracellular domain of mouse E-Cadherin, a 120-kDa transmembrane cell-cell adhesion glycoprotein also known as CD324 and uvomorulin. E-Cadherin is a critical regulator of epithelial junction formation and is involved in development, bacterial pathogenesis, and tumor invasion.
- Reactivity: Mouse E-Cadherin
- Isotype: Rat IgG1, κ
- Immunogen: Mouse PCC4azal teratocarcinoma cells
- Purity: ≥95% determined by SDS-PAGE
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Reported applications: In vivo E-Cadherin neutralization, in vitro E-Cadherin neutralization, immunofluorescence, immunoprecipitation, western blot
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.