Learn More
Bio X Cell InVivoMAb Antibody, E-Cadherin, Clone DECMA-1, 50 MG
Supplier: Bio X Cell BE0352R050MG
The DECMA-1 monoclonal antibody reacts with the extracellular domain of mouse E-Cadherin (CD324), a 120-kDa transmembrane cell-cell adhesion glycoprotein. It regulates epithelial junction formation and is involved in development, bacterial pathogenesis, and tumor invasion. This antibody can delay tumor onset and attenuate tumor burden.
- Reactivity: mouse
- Isotype: rat IgG1, κ
- Immunogen: mouse PCC4azal teratocarcinoma cells
- Formulation: PBS, pH 7.0
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Production: purified from cell culture supernatant, animal-free facility
- Purification: protein G
- Molecular weight: 150 kDa
- Applications: in vivo and in vitro E-Cadherin neutralization, immunofluorescence, immunoprecipitation, Western blot
- Storage: store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.