Learn More
Bio X Cell Isotype Control, Mouse IgG2c, Monoclonal, Anti-Dengue Virus, 1 MG
Supplier: Bio X Cell BE0366R001MG
This monoclonal antibody serves as an isotype-matched control for mouse IgG2c antibodies. It is suitable for a variety of in vivo and in vitro applications, with the exception of those involving dengue virus. Purified from cell culture supernatant, it is formulated in PBS at pH 7.0.
- Isotype: Mouse IgG2c,
- Clone: DV5-1
- Unconjugated format
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin level: 1 EU/mg
- Purity: 95%
- Sterility: 0.2 m filtration
- Produced in an animal-free facility
- Purified by Protein A
- Molecular weight: 150 kDa
- Store at 4C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.