Learn More
Bio X Cell InVivoMAb Isotype Control, Mouse IgG2c, DV5-1, 25 MG, 25 MG
Supplier: Bio X Cell BE0366R025MG
This InVivoMAb mouse IgG2c isotype control, clone DV5-1, is designed to react with dengue virus. It serves as an isotype-matched control for mouse IgG2c antibodies in both in vivo and in vitro applications, excluding those involving dengue virus. Formulated in PBS at pH 7.0 without stabilizers or preservatives, it is purified from cell culture supernatant in an animal-free facility using Protein A.
- Isotype: Mouse IgG2c,
- Recommended dilution buffer: pH 7.0 Dilution Buffer
- Unconjugated (conjugation services available)
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin level: 1EU/mg
- Purity: 95% by SDS-PAGE
- Sterility: 0.2 m filtration
- Production: Animal-free facility
- Purification method: Protein A
- Molecular weight: 150 kDa
- Storage: 4C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.