Learn More
Bio X Cell Isotype Control, DV5-1, Anti-Dengue Virus, 50 MG
Supplier: Bio X Cell BE0366R050MG
The DV5-1 monoclonal antibody reacts with dengue virus. This antibody is ideal for use as an isotype-matched control for mouse IgG2c antibodies in in vivo and in vitro applications, excluding those involving dengue virus.
- Clone DV5-1
- Purity ≥95% by SDS-PAGE
- Endotoxin ≤1EU/mg (≤0.001EU/µg) by LAL assay
- Sterile filtered to 0.2 µm
- Purified by protein A from animal-free facility cell culture supernatant
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Molecular weight 150 kDa
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.